Skip to Content
Merck

AABH97CD8293

AA BLOCKS, INC.

Diazoxide-d3

Synonym(s):

7-chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide d_3_

Select a Size

Change View

About This Item

Linear Formula:
C8H7O2N2Cl1S1
CAS Number:
Molecular Weight:
232.69
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H7ClN2O2S/c1-5-10-7-3-2-6(9)4-8(7)14(12,13)11-5/h2-4,10-11H,1H2/i1D2

Inchi Key

LOKZAVCIAGCAKR-DICFDUPASA-N

Country of Origin

US

Purity

95%

Storage temp.

2~8C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library