Skip to Content
Merck

AABH97CD82FE

AA BLOCKS, INC.

Eprinomectin B1a

Synonym(s):

N-((2S,3R,4S,6S)-6-(((2S,3S,4S,6R)-6-(((2S,3'S,5S,6R,7'R,9'Z,11'S,12'S,13'Z,15'Z,15a1'S,17a'R,18'R,20a'R)-6-((S)-sec-butyl)-15a1',18'-dihydroxy-5,10',12',19'-tetramethyl-1'-oxo-3',4',5,6,8',11',12',15a1',16',17a',18',20a'-dodecahydro-1'H,7'H-spiro[pyran-2,5'-[3,7]methano[1,5]dioxacyclooctadecino[9,8,7-cd]benzofuran]-11'-yl)oxy)-4-methoxy-2-methyltetrahydro-2H-pyran-3-yl)oxy)-4-methoxy-2-methyltetrahydro-2H-pyran-3-yl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C50H75NO14/c1-12-26(2)45-29(5)18-19-49(65-45)24-36-21-35(64-49)17-16-28(4)44(27(3)14-13-15-34-25-58-47-43(53)30(6)20-37(48(54)61-36)50(34,47)55)62-41-23-39(57-11)46(32(8)60-41)63-40-22-38(56-10)42(31(7)59-40)51-33(9)52/h13-16,18-20,26-27,29,31-32,35-47,53,55H,12,17,21-25H2,1-11H3,(H,51,52)/b14-13-,28-16-,34-15-/t26-,27-,29-,31-,32-,35+,36-,37-,38-,39-,40-,41-,42+,43+,44-,45+,46-,47+,49+,50+/m0/s1

Inchi Key

ZKWQQXZUCOBISE-HENTUUONSA-N

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library