Skip to Content
Merck

AABH97CD85B0

AA BLOCKS, INC.

Prodigiosin

Synonym(s):

(E)-4-methoxy-5-((5-methylene-4-pentyl-1,5-dihydro-2H-pyrrol-2-ylidene)methyl)-1H,1'H-2,2'-bipyrrole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H25O1N3
CAS Number:
Molecular Weight:
323.44
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H25N3O/c1-4-5-6-8-15-11-16(22-14(15)2)12-19-20(24-3)13-18(23-19)17-9-7-10-21-17/h7,9-13,21-23H,2,4-6,8H2,1,3H3/b16-12+

Inchi Key

SHJCXLRWNZWNJE-FOWTUZBSSA-N

Purity

98%

Storage temp.

-20C

Country of Origin

US



Still not finding the right product?

Explore all of our products under Prodigiosin




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library