Skip to Content
Merck

AABH97CD863D

AA BLOCKS, INC.

Flupirtine-d4 (hydrochloride)

Synonym(s):

ethyl 2-amino-6-[(4-fluorobenzyl)amino]-3-pyridinylcarbamate hydrochloride d_4_

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H18O2N4Cl1F1
CAS Number:
Molecular Weight:
340.78
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H17FN4O2.ClH/c1-2-22-15(21)19-12-7-8-13(20-14(12)17)18-9-10-3-5-11(16)6-4-10;/h3-8H,2,9H2,1H3,(H,19,21)(H3,17,18,20);1H/i3D,4D,5D,6D;

Inchi Key

WBAZQELGBFQHPE-HGSONKNXSA-N

Purity

99%14

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library