Skip to Content
Merck

AABH97CD867E

AA BLOCKS, INC.

Sulprostone

A solution in methyl acetate

Synonym(s):

(5E)-7-((1R,2R,3R)-3-hydroxy-2-((R,E)-3-hydroxy-4-phenoxybut-1-en-1-yl)-5-oxocyclopentyl)-N-(methylsulfonyl)hept-5-enimidic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Purity

95%

Country of Origin

US

Inchi Code

1S/C23H31NO7S/c1-32(29,30)24-23(28)12-8-3-2-7-11-19-20(22(27)15-21(19)26)14-13-17(25)16-31-18-9-5-4-6-10-18/h2,4-7,9-10,13-14,17,19-20,22,25,27H,3,8,11-12,15-16H2,1H3,(H,24,28)/b7-2+,14-13+/t17-,19-,20-,22-/m1/s1

Inchi Key

UQZVCDCIMBLVNR-MCOQOZQRSA-N

Storage temp.

-20C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library