Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C22H18N4O2S/c1-23-22(27)18-7-2-3-8-21(18)29(28)16-10-11-17-19(25-26-20(17)14-16)12-9-15-6-4-5-13-24-15/h2-14H,1H3,(H,23,27)(H,25,26)/b12-9+
Inchi Key
MAHSNFBPPYMWFB-FMIVXFBMSA-N
Purity
98%
Country of Origin
US
Storage temp.
2~8C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.