Skip to Content
Merck

Skip To

AABH97CD8A21

AA BLOCKS, INC.

(2S)-2-Amino-4-((2-amino-2-carboxyethyl)thio)butanoic acid

Synonym(s):

(2S)-2-amino-4-(2-amino-2-carboxyethylthio)butanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C7H14O4N2S1
CAS Number:
Molecular Weight:
222.27
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H14N2O4S/c8-4(6(10)11)1-2-14-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)/t4-,5?/m0/s1

Inchi Key

ILRYLPWNYFXEMH-ROLXFIACSA-N

Purity

90%

Storage temp.

2~8C

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library