Storage temp.
N/A
Inchi Code
1S/Ca.ClHO4.2H/c;2-1(3,4)5;;/h;(H,2,3,4,5);;
Inchi Key
QHNMGOPAZNDLNL-UHFFFAOYSA-N
Purity
99%
Country of Origin
US
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.