Skip to Content
Merck

AABH97CDA3DE

AA BLOCKS, INC.

Gamma-glutamylcysteine

Synonym(s):

(2S)-2-amino-5-{[(1R)-1-carboxy-2-sulfanylethyl]amino}-5-oxopentanoic acid

Select a Size

Change View

About This Item

Linear Formula:
C8H14O5N2S1
CAS Number:
Molecular Weight:
250.28
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H14N2O5S/c9-4(7(12)13)1-2-6(11)10-5(3-16)8(14)15/h4-5,16H,1-3,9H2,(H,10,11)(H,12,13)(H,14,15)/t4-,5-/m0/s1

Inchi Key

RITKHVBHSGLULN-WHFBIAKZSA-N

Storage temp.

N/A

Country of Origin

US



Still not finding the right product?

Explore all of our products under Gamma-glutamylcysteine




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library