Skip to Content
Merck

AABH97D2AD8A

AA BLOCKS, INC.

L-Ornithine,N5-(4,5-dihydro-4-methyl-5-oxo-1H-imidazol-2-yl)-

Synonym(s):

(2S)-2-amino-5-[(5-methyl-4-oxo-4,5-dihydro-1H-imidazol-2-yl)amino]pentanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H16O3N4
CAS Number:
Molecular Weight:
228.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H16N4O3/c1-5-7(14)13-9(12-5)11-4-2-3-6(10)8(15)16/h6,14H,2-4,10H2,1H3,(H,15,16)(H2,11,12,13)/t6-/m0/s1

Inchi Key

SGYDNAPOJZORFJ-LURJTMIESA-N

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library