Skip to Content
Merck

AABH97D2C4EF

AA BLOCKS, INC.

N-Ethyl Fluprostenol Carboxamide

Synonym(s):

(5E)-7-((1R,2R,3R,5S)-3,5-dihydroxy-2-((R,E)-3-hydroxy-4-(3-(trifluoromethyl)phenoxy)but-1-en-1-yl)cyclopentyl)-N-ethylhept-5-enimidic acid

Select a Size

Change View

About This Item

Linear Formula:
C25H34O5N1F3
CAS Number:
Molecular Weight:
485.54
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Country of Origin

US

Inchi Code

1S/C25H34F3NO5/c1-2-29-24(33)11-6-4-3-5-10-20-21(23(32)15-22(20)31)13-12-18(30)16-34-19-9-7-8-17(14-19)25(26,27)28/h3,5,7-9,12-14,18,20-23,30-32H,2,4,6,10-11,15-16H2,1H3,(H,29,33)/b5-3+,13-12+/t18-,20-,21-,22+,23-/m1/s1

Inchi Key

GAAVJEMZITUZOC-NIQHAARNSA-N

Storage temp.

-20C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library