Skip to Content
Merck

AABH97D2CEB5

AA BLOCKS, INC.

butyl 2-aminobenzoate

Synonym(s):

butyl 2-aminobenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H15O2N1
CAS Number:
Molecular Weight:
193.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H15NO2/c1-2-3-8-14-11(13)9-6-4-5-7-10(9)12/h4-7H,2-3,8,12H2,1H3

Inchi Key

HUIYGGQINIVDNW-UHFFFAOYSA-N

Purity

98100%

Storage temp.

RT

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library