Skip to Content
Merck

AABH97D2D3FB

AA BLOCKS, INC.

Pterosin A

Synonym(s):

(S)-6-(2-hydroxyethyl)-2-(hydroxymethyl)-2,5,7-trimethyl-2,3-dihydro-1H-inden-1-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H20O3
CAS Number:
Molecular Weight:
248.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H20O3/c1-9-6-11-7-15(3,8-17)14(18)13(11)10(2)12(9)4-5-16/h6,16-17H,4-5,7-8H2,1-3H3/t15-/m0/s1

Inchi Key

BDZJLPDYMKPKGC-HNNXBMFYSA-N

Purity

98%

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library