Skip to Content
Merck

AABH97D2EFE0

AA BLOCKS, INC.

Cyclohexanemethanol, 4-pentyl-, trans-

Synonym(s):

(4-pentylcyclohexyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H24O1
CAS Number:
Molecular Weight:
184.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H24O/c1-2-3-4-5-11-6-8-12(10-13)9-7-11/h11-13H,2-10H2,1H3/t11-,12-

Inchi Key

RAOQLAYIDNKQHG-HAQNSBGRSA-N

Purity

95%

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library