Skip to Content
Merck

AABH97D2F08E

AA BLOCKS, INC.

Indoramin

Synonym(s):

N-{1-[2-(1H-indol-3-yl)ethyl]-4-piperidinyl}benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C22H25O1N3
CAS Number:
Molecular Weight:
347.46
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C22H25N3O/c26-22(17-6-2-1-3-7-17)24-19-11-14-25(15-12-19)13-10-18-16-23-21-9-5-4-8-20(18)21/h1-9,16,19,23H,10-15H2,(H,24,26)

Inchi Key

JXZZEXZZKAWDSP-UHFFFAOYSA-N

Purity

98%

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library