Skip to Content
Merck

AABH97D3DB0E

AA BLOCKS, INC.

4-hydroxy Omeprazole sulfide

Synonym(s):

2-(((6-methoxy-1H-benzo[d]imidazol-2-yl)thio)methyl)-3,5-dimethylpyridin-4-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C16H17N3O2S/c1-9-7-17-14(10(2)15(9)20)8-22-16-18-12-5-4-11(21-3)6-13(12)19-16/h4-7H,8H2,1-3H3,(H,17,20)(H,18,19)

Inchi Key

LVUFHVGKGMRSQW-UHFFFAOYSA-N

Purity

98%

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library