Skip to Content
Merck

AABH99A44F2F

AA BLOCKS, INC.

Loxapine-d8 Hydrochloride

Synonym(s):

2-chloro-11-(4-methylpiperazin-1-yl-2,2,3,3,5,5,6,6-d8)dibenzo[b,f][1,4]oxazepine hydrochloride

Select a Size

Change View

About This Item

Linear Formula:
C18H19O1N3Cl2
CAS Number:
Molecular Weight:
372.32
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C18H18ClN3O.ClH/c1-21-8-10-22(11-9-21)18-14-12-13(19)6-7-16(14)23-17-5-3-2-4-15(17)20-18;/h2-7,12H,8-11H2,1H3;1H/i8D2,9D2,10D2,11D2;

Inchi Key

JSXBVMKACNEMKY-KTSBLNPMSA-N

Country of Origin

US

Storage temp.

-20C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library