Skip to Content
Merck

AABH99A4B2ED

AA BLOCKS, INC.

Caerulein (acetate)

Synonym(s):

(3S,6S,9S,15S,18S)-9-((1H-indol-3-yl)methyl)-3-((1-amino-1-oxo-3-phenylpropan-2-yl)carbamoyl)-21-((S)-5-amino-5-oxo-2-((S)-5-oxopyrrolidine-2-carboxamido)pentanamido)-15-((R)-1-hydroxyethyl)-6-(2-(methylthio)ethyl)-5,8,11,14,17,20-hexaoxo-18-(4-(sulfooxy)benzyl)-4,7,10,13,16,19-hexaazatricosanedioic acid--acetic acid (1/1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C58H73N13O21S2.C2H4O2/c1-29(72)49(71-57(87)40(23-31-12-14-33(15-13-31)92-94(89,90)91)68-56(86)43(26-48(78)79)69-52(82)37(16-18-44(59)73)65-51(81)36-17-19-45(74)63-36)58(88)62-28-46(75)64-41(24-32-27-61-35-11-7-6-10-34(32)35)54(84)66-38(20-21-93-2)53(83)70-42(25-47(76)77)55(85)67-39(50(60)80)22-30-8-4-3-5-9-30;1-2(3)4/h3-15,27,29,36-43,49,61,72H,16-26,28H2,1-2H3,(H2,59,73)(H2,60,80)(H,62,88)(H,63,74)(H,64,75)(H,65,81)(H,66,84)(H,67,85)(H,68,86)(H,69,82)(H,70,83)(H,71,87)(H,76,77)(H,78,79)(H,89,90,91);1H3,(H,3,4)/t29-,36+,37+,38+,39+,40+,41+,42+,43+,49+;/m1./s1

Inchi Key

BMJCGMRTEFIGQJ-CBCFNHQSSA-N

Purity

95%

Storage temp.

N/A

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library