Skip to Content
Merck

B68720

5-Bromoindole-3-acetic acid

97%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H8BrNO2
CAS Number:
Molecular Weight:
254.08
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
254-917-8
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

97%

mp

143-145 °C (lit.)

SMILES string

OC(=O)Cc1c[nH]c2ccc(Br)cc12

InChI

1S/C10H8BrNO2/c11-7-1-2-9-8(4-7)6(5-12-9)3-10(13)14/h1-2,4-5,12H,3H2,(H,13,14)

InChI key

WTFGHMZUJMRWBK-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library