Skip to Content
Merck

GF09831720

Phosphor bronze alloy, Cu94Sn6

foil, 8mm disks, 0.076mm thickness, as rolled

Synonym(s):

CU050245

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.23
UNSPSC Code:
11101711
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

foil (disks - as rolled)

composition

Cu94Sn6

reaction suitability

core: copper, core: tin

manufacturer/tradename

Goodfellow 098-317-20

size × thickness

8 mm × 0.076 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow


Still not finding the right product?

Explore all of our products under Phosphor bronze alloy, Cu94Sn6




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library