Skip to Content

GF80348564

Zinc

lump, 2 mm max. lump size, weight 1000 g, purity 99.98%

Synonym(s):

ZN006100

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Zn
CAS Number:
Molecular Weight:
65.39
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141750
MDL number:
eCl@ss:
38150401
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

1 mmHg ( 487 °C)

assay

99.98%

form

lumps

manufacturer/tradename

Goodfellow 803-485-64

resistivity

5.8 μΩ-cm, 20°C

weight

1000 g

max. lump size

2 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

application(s)

electroplating

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow


Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library