Skip to Content
Merck

Skip To

GF96886355

Alumina

bolt, size m6 x 10 mm, length 25 pcs, head style hexagonal

Synonym(s):

AL60BO61

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
32 600,00 CZK

About This Item

NACRES:
NA.23
UNSPSC Code:
31161620

32 600,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 25

manufacturer/tradename

Goodfellow 968-863-55

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GF43225860GF26996833
description

bolt, size m6 x 10 mm, length 25 pcs, head style hexagonal

description

Bolt, size m6 x 10 mm, length 10 pcs, head style hexagonal

description

rod, 5.0 mm diameter, length 50 mm, purity 99%

manufacturer/tradename

Goodfellow 968-863-55

manufacturer/tradename

Goodfellow 432-258-60

manufacturer/tradename

Goodfellow 269-968-33

packaging

pkg of 25

packaging

pkg of 10

packaging

-


Still not finding the right product?

Explore all of our products under Alumina




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters