Skip to Content
Merck

Skip To

PH018564

tert-Butyl (1R,5S)-3-[(methylsulfonyl)oxy]-8-azabicyclo[3.2.1]octane-8-carboxylate

AldrichCPR

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
C13H23O5N1S1
UNSPSC Code:
12352200
MDL number:


form

solid

SMILES string

O=S(OC1C[C@H]2N(C(OC(C)(C)C)=O)[C@H](CC2)C1)(C)=O

InChI

1S/C13H23NO5S/c1-13(2,3)18-12(15)14-9-5-6-10(14)8-11(7-9)19-20(4,16)17/h9-11H,5-8H2,1-4H3/t9-,10+,11+

InChI key

YLTJZCYZECLGGN-URLYPYJESA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters