Skip to Content
Merck

ATEH98884792

AstaTech, Inc.

FLUVALINATE

Synonym(s):

cyano(3-phenoxyphenyl)methyl (2R)-2-[2-chloro-4-(trifluoromethyl)anilino]-3-methylbutanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C26H22O3N2Cl1F3
CAS Number:
Molecular Weight:
502.92
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C26H22ClF3N2O3/c1-16(2)24(32-22-12-11-18(14-21(22)27)26(28,29)30)25(33)35-23(15-31)17-7-6-10-20(13-17)34-19-8-4-3-5-9-19/h3-14,16,23-24,32H,1-2H3/t23?,24-/m1/s1

Inchi Key

INISTDXBRIBGOC-XMMISQBUSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Room temperature

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?

Explore all of our products under FLUVALINATE




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library