Skip to Content

BDPH9BCC1998

Bide Pharmatech

2-Bromo-1-(2,6-difluorophenyl)ethanone

Synonym(s):

2-bromo-1-(2,6-difluorophenyl)ethanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H5O1Br1F2
CAS Number:
Molecular Weight:
235.03
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H5BrF2O/c9-4-7(12)8-5(10)2-1-3-6(8)11/h1-3H,4H2

Inchi Key

UZKLFNHMBFDXEN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library