Skip to Content
Merck

BDPH9BCC2733

Bide Pharmatech

2,6-Difluoro-3-(trifluoromethyl)benzoic acid

Synonym(s):

2,6-difluoro-3-(trifluoromethyl)benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H3O2F5
CAS Number:
Molecular Weight:
226.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H3F5O2/c9-4-2-1-3(8(11,12)13)6(10)5(4)7(14)15/h1-2H,(H,14,15)

Inchi Key

CVJDKSBPBBKDEN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library