Skip to Content

BDPH9BCC3D24

Bide Pharmatech

4-Iodo-3-(trifluoromethoxy)aniline

Synonym(s):

4-iodo-3-(trifluoromethoxy)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5O1N1F3I1
CAS Number:
Molecular Weight:
303.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H5F3INO/c8-7(9,10)13-6-3-4(12)1-2-5(6)11/h1-3H,12H2

Inchi Key

VDVMGAPDTJTWDH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid or solid



Still not finding the right product?

Explore all of our products under 4-Iodo-3-(trifluoromethoxy)aniline




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library