Skip to Content
Merck

BDPH9BCC3D2C

Bide Pharmatech

4-(5-(4-Fluorophenyl)-1,3,4-oxadiazol-2-yl)piperidine

Synonym(s):

4-[5-(4-fluorophenyl)-1,3,4-oxadiazol-2-yl]piperidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H14O1N3F1
CAS Number:
Molecular Weight:
247.27
MDL number:
Product BDPH9BCC3D2C is not currently sold in your country.


Inchi Code

1S/C13H14FN3O/c14-11-3-1-9(2-4-11)12-16-17-13(18-12)10-5-7-15-8-6-10/h1-4,10,15H,5-8H2

Inchi Key

OCVSOAJAJMMNGN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library