Skip to Content
Merck

BDPH9BCC42E1

Bide Pharmatech

1-(4-Chlorophenyl)-1H-imidazole-4-carboxylic acid

Synonym(s):

1-(4-chlorophenyl)-1H-imidazole-4-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H7O2N2Cl1
CAS Number:
Molecular Weight:
222.63
MDL number:
Product BDPH9BCC42E1 is not currently sold in your country.


Inchi Code

1S/C10H7ClN2O2/c11-7-1-3-8(4-2-7)13-5-9(10(14)15)12-6-13/h1-6H,(H,14,15)

Inchi Key

LDALVFKCZXOZGI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library