Skip to Content
Merck

BDPH9BCC5582

Bide Pharmatech

4-[(2,4-Dimethylphenyl)sulfanyl]aniline hydrochloride

Synonym(s):

4-[(2,4-dimethylphenyl)sulfanyl]aniline hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H16N1Cl1S1
CAS Number:
Molecular Weight:
265.81
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid

Inchi Code

1S/C14H15NS.ClH/c1-10-3-8-14(11(2)9-10)16-13-6-4-12(15)5-7-13;/h3-9H,15H2,1-2H3;1H

Inchi Key

DLQYXWSCPPGOJA-UHFFFAOYSA-N



Still not finding the right product?


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library