Skip to Content
Merck

BDPH9BCC592E

Bide Pharmatech

3-Cyclohexyl-1H-indole-6-carboxylic acid

Synonym(s):

3-cyclohexyl-1H-indole-6-carboxylic acid

Select a Size

Change View

About This Item

Linear Formula:
C15H17O2N1
CAS Number:
Molecular Weight:
243.31
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H17NO2/c17-15(18)11-6-7-12-13(9-16-14(12)8-11)10-4-2-1-3-5-10/h6-10,16H,1-5H2,(H,17,18)

Inchi Key

OZJSQVKDKOOQIT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to light-yellow powder or crystals



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library