Skip to Content
Merck

BDPH9BCC6C20

Bide Pharmatech

tert-Butyl (4-chlorobutyl)carbamate

Synonym(s):

tert-butyl 4-chlorobutylcarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H18O2N1Cl1
CAS Number:
Molecular Weight:
207.7
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H18ClNO2/c1-9(2,3)13-8(12)11-7-5-4-6-10/h4-7H2,1-3H3,(H,11,12)

Inchi Key

BEKDZRXUWYOOPU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

White to Yellow Solid or Semi-solid or liquid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library