Skip to Content
Merck

BDPH9BCC76DA

Bide Pharmatech

tert-Butyl 4-(bromomethyl)-1H-benzo[d]imidazole-1-carboxylate

Synonym(s):

tert-butyl 4-(bromomethyl)-1H-benzimidazole-1-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H15O2N2Br1
CAS Number:
Molecular Weight:
311.18
MDL number:
Product BDPH9BCC76DA is not currently sold in your country.


Inchi Code

1S/C13H15BrN2O2/c1-13(2,3)18-12(17)16-8-15-11-9(7-14)5-4-6-10(11)16/h4-6,8H,7H2,1-3H3

Inchi Key

RJQPRKXEVWZMAO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to yellow powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library