Skip to Content
Merck

BDPH9BCC7966

Bide Pharmatech

1,3-Dithian-5-amine

Synonym(s):

1,3-dithian-5-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H9N1S2
CAS Number:
Molecular Weight:
135.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H9NS2/c5-4-1-6-3-7-2-4/h4H,1-3,5H2

Inchi Key

FPYHBGHCORESFC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 1,3-Dithian-5-amine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library