Skip to Content
Merck

BDPH9BCC7E93

Bide Pharmatech

4-[2-(4-Bromophenoxy)ethyl]morpholine

Synonym(s):

4-[2-(4-bromophenoxy)ethyl]morpholine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H16O2N1Br1
CAS Number:
Molecular Weight:
286.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H16BrNO2/c13-11-1-3-12(4-2-11)16-10-7-14-5-8-15-9-6-14/h1-4H,5-10H2

Inchi Key

SWXZUUKQIQPHIC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid or semi-solid or solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library