Skip to Content

BDPH9BCC8DA9

Bide Pharmatech

N-(4-Formylpyridin-3-yl)acetamide

Synonym(s):

N-(4-formyl-3-pyridinyl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H8O2N2
CAS Number:
Molecular Weight:
164.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H8N2O2/c1-6(12)10-8-4-9-3-2-7(8)5-11/h2-5H,1H3,(H,10,12)

Inchi Key

QXEFMYBPXJVIHG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Light yellow to yellow brown powder or crystals



Still not finding the right product?

Explore all of our products under N-(4-Formylpyridin-3-yl)acetamide


hstatements

signalword

Danger

pictograms

Skull and crossbones


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library