Skip to Content

BDPH9BCC8F4F

Bide Pharmatech

2-(Chloromethyl)-5-(trifluoromethyl)pyrazine

Synonym(s):

2-(chloromethyl)-5-(trifluoromethyl)pyrazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H4N2Cl1F3
CAS Number:
Molecular Weight:
196.56
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H4ClF3N2/c7-1-4-2-12-5(3-11-4)6(8,9)10/h2-3H,1H2

Inchi Key

FDDCRYZDGUBGAZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library