Product BDPH9BCCAA2E is not currently sold in your country.
Inchi Code
1S/C9H6ClNO3S/c10-15(12,13)8-3-1-7(2-4-8)9-5-11-6-14-9/h1-6H
Inchi Key
QCGUTLAPXWBIMM-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Explore all of our products under 4-(Oxazol-5-yl)benzene-1-sulfonyl chloride
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

