Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C9H8F2O/c1-2-9(12)6-3-7(10)5-8(11)4-6/h3-5H,2H2,1H3
Inchi Key
FVDQWXARVQADKN-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Liquid or low-melting solid
Still not finding the right product?
Explore all of our products under 1-(3,5-Difluorophenyl)propan-1-one
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
