Skip to Content
Merck

BDPH9BCCBDBD

Bide Pharmatech

4-Methoxy-2-methylaniline

Synonym(s):

4-methoxy-2-methylaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H11O1N1
CAS Number:
Molecular Weight:
137.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H11NO/c1-6-5-7(10-2)3-4-8(6)9/h3-5H,9H2,1-2H3

Inchi Key

CDGNLUSBENXDGG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Yellow-brown or red-brown to black or purple liquid or solid

Melting Point

13-14C



Still not finding the right product?

Explore all of our products under 4-Methoxy-2-methylaniline



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library