Skip to Content
Merck

BDPH9BCCBDF4

Bide Pharmatech

2-Bromo-3-isopropylpyridine

Synonym(s):

2-bromo-3-isopropylpyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H10N1Br1
CAS Number:
Molecular Weight:
200.08
MDL number:
Product BDPH9BCCBDF4 is not currently sold in your country.


Inchi Code

1S/C8H10BrN/c1-6(2)7-4-3-5-10-8(7)9/h3-6H,1-2H3

Inchi Key

JEBINPKMSQABNA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to yellow liquid



Still not finding the right product?

Explore all of our products under 2-Bromo-3-isopropylpyridine


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library