Skip to Content
Merck

BDPH9BCCBF09

Bide Pharmatech

3,5-Dibromoaniline

Synonym(s):

3,5-dibromoaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5N1Br2
CAS Number:
Molecular Weight:
250.92
MDL number:
Product BDPH9BCCBF09 is not currently sold in your country.


Inchi Code

1S/C6H5Br2N/c7-4-1-5(8)3-6(9)2-4/h1-3H,9H2

Inchi Key

RVNUUWJGSOHMRR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Pale-yellow to brown to black or gray powder or crystals



Still not finding the right product?

Explore all of our products under 3,5-Dibromoaniline


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library