Skip to Content
Merck

BDPH9BCCBFB8

Bide Pharmatech

Cyclopropyl(4-hydroxyphenyl)methanone

Synonym(s):

cyclopropyl(4-hydroxyphenyl)methanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H10O2
CAS Number:
Molecular Weight:
162.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H10O2/c11-9-5-3-8(4-6-9)10(12)7-1-2-7/h3-7,11H,1-2H2

Inchi Key

GVBHKNVCIKUPCZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Yellow to Yellow-brown Solid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library