Skip to Content
Merck

BDPH9BCCC8E7

Bide Pharmatech

1-(3-(Difluoromethyl)-1-methyl-1H-pyrazol-4-yl)ethanone

Synonym(s):

1-(3-(difluoromethyl)-1-methyl-1H-pyrazol-4-yl)ethan-1-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H8O1N2F2
CAS Number:
Molecular Weight:
174.15
MDL number:
Product BDPH9BCCC8E7 is not currently sold in your country.


Inchi Code

1S/C7H8F2N2O/c1-4(12)5-3-11(2)10-6(5)7(8)9/h3,7H,1-2H3

Inchi Key

BEZVCXLDJHKSQE-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

97%

Country of Origin

CN

Physical Form

Solid or liquid

Storage temp.

Inert atmosphere, store in freezer, under -20°C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library