Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C8H12O3/c9-7-2-1-3-8(6-7)10-4-5-11-8/h1-6H2
Inchi Key
TVXYCOHQUCHFAM-UHFFFAOYSA-N
Shipping Temperature
Cold-chain transportation
Purity
97%
Storage temp.
Inert atmosphere, store in freezer, under -20C
Country of Origin
CN
Physical Form
Colorless to Yellow Liquid
Still not finding the right product?
Explore all of our products under 1,4-Dioxaspiro[4.5]decan-7-one
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
