Skip to Content
Merck

BDPH9BCCCC3E

Bide Pharmatech

2,7-Dibromoquinoline

Synonym(s):

2,7-dibromoquinoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H5N1Br2
CAS Number:
Molecular Weight:
286.95
MDL number:
Product BDPH9BCCCC3E is not currently sold in your country.


Inchi Code

1S/C9H5Br2N/c10-7-3-1-6-2-4-9(11)12-8(6)5-7/h1-5H

Inchi Key

IDHADAAFHVDGIL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid



Still not finding the right product?

Explore all of our products under 2,7-Dibromoquinoline



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library