Skip to Content
Merck

BDPH9BCCCE14

Bide Pharmatech

Spiro[3.3]heptane-2-carboxylic acid

Synonym(s):

spiro[3.3]heptane-2-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H12O2
CAS Number:
Molecular Weight:
140.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H12O2/c9-7(10)6-4-8(5-6)2-1-3-8/h6H,1-5H2,(H,9,10)

Inchi Key

FUQHLUSGMOSPRQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library