Skip to Content
Merck

Skip To

BDPH9BCCD522

Bide Pharmatech

5-Chloro-2-(methylthio)-4-(trifluoromethyl)pyrimidine

Synonym(s):

5-chloro-2-(methylthio)-4-(trifluoromethyl)pyrimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H4N2Cl1S1F3
CAS Number:
Molecular Weight:
228.63
MDL number:
Product BDPH9BCCD522 is not currently sold in your country.


Inchi Code

1S/C6H4ClF3N2S/c1-13-5-11-2-3(7)4(12-5)6(8,9)10/h2H,1H3

Inchi Key

IRPJOYRKSXBYBE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid



Still not finding the right product?


hstatements

signalword

Danger

pictograms

Skull and crossbones


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library