Skip to Content
Merck

BDPH9BCCD6BD

Bide Pharmatech

2-Bromo-5-methoxypyridin-4-amine

Synonym(s):

2-bromo-5-methoxypyridin-4-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H7O1N2Br1
CAS Number:
Molecular Weight:
203.04
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H7BrN2O/c1-10-5-3-9-6(7)2-4(5)8/h2-3H,1H3,(H2,8,9)

Inchi Key

ZMXFLZUNHINXLA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-Bromo-5-methoxypyridin-4-amine


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library