Skip to Content
Merck

BDPH9BCCDECD

Bide Pharmatech

4-Methoxypicolinic acid

Synonym(s):

4-methoxy-2-pyridinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7O3N1
CAS Number:
Molecular Weight:
153.14
MDL number:
Product BDPH9BCCDECD is not currently sold in your country.


Inchi Code

1S/C7H7NO3/c1-11-5-2-3-8-6(4-5)7(9)10/h2-4H,1H3,(H,9,10)

Inchi Key

UELRAKDBDJRXST-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: solid



Still not finding the right product?

Explore all of our products under 4-Methoxypicolinic acid


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library